Learn More
Mercodia Inc Ultrasensitive Mouse Insulin ELISA, Mouse, Serum Plasma, 96 Wells
SDP
Supplier:  Mercodia Inc 10124901
Encompass_Preferred
Mercodia Ultrasensitive Mouse Insulin ELISA provides a method for the quantitative determination of insulin in mouse serum, plasma, or cell culture media. The Mercodia Ultrasensitive Mouse Insulin ELISA is an easy-to-use assay based on highly specific monoclonal antibodies with insignificant or no cross-reactivity to C-peptide or proinsulin. Offers low measuring range.
Catalog No. NC0035732