Learn More
Alkali Scientific Ultra Low Attachment Dish, 100mm with external grip edge, 10/Cs
SDP
Catalog No. 502335808
Encompass_Preferred
Ultra Low Attachment (ULA) Cell Tissue Culture Dishes▪ Ultra Low Cell Adhesion▪ Comprised of a covalently bound hydrogel layer that minimizes cell attachment, protein absorption & cellular activation.▪ Use when culturing stem cells and regenerative cell therapy▪ Promotes growth of suspension cell lines - both established and primary cells▪ Ideal environment to help nurture ebryoid body formation from ES cells▪ Flat Bottom▪ Product Shelf Life: 3 years at room temperature storage (25 ℃)
Catalog No. 50-233-5808 Supplier Alkali Scientific Supplier No. CFD100
Ultra Low Attachment (ULA) Cell Tissue Culture Dishes▪ Ultra Low Cell Adhesion▪ Comprised of a covalently bound hydrogel layer that minimizes cell attachment, protein absorption & cellular activation.▪ Use when culturing stem cells and regenerative cell therapy▪ Promotes growth of suspension cell lines - both established and primary cells▪ Ideal environment to help nurture ebryoid body formation from ES cells▪ Flat Bottom▪ Product Shelf Life: 3 years at room temperature storage (25 ℃)