Learn More
Alkali Scientific UltraSOFT 5000™ Powder Free Nitrile Glove, Extra-Large, 250/box, 2500/cs
SDP
Catalog No. 502335922
Encompass_Preferred
Three times more per case. Highly elastic comfortable feel. Powder-free with textured finger tips. 250 gloves per box, 2500 gloves per case. Thickness: Cuff: 2.4 mil Palm: 2.4 mil Fingers: 3.1 mil Strength: Tensile on spec sheet: 23 Mpa Tensile Typical Value: 32 Mp
Catalog No. 50-233-5922 Supplier Alkali Scientific Supplier No. NG2225
Three times more per case. Highly elastic comfortable feel. Powder-free with textured finger tips. 250 gloves per box, 2500 gloves per case. Thickness: Cuff: 2.4 mil Palm: 2.4 mil Fingers: 3.1 mil Strength: Tensile on spec sheet: 23 Mpa Tensile Typical Value: 32 Mp