Learn More
Alkali Scientific UltraSOFT 5000™ Powder Free Nitrile Glove, Extra-Small, 300/box, 3000/cs
SDP
Catalog No. 502335882
Encompass_Preferred
Three times more per case. Highly elastic comfortable feel. Powder-free with textured finger tips. 300 gloves per box, 3000 gloves per case. Thickness: Cuff: 2.4 mil Palm: 2.4 mil Fingers: 3.1 mil Strength: Tensile on spec sheet: 23 Mpa Tensile Typical Value: 32 Mp
Catalog No. 50-233-5882 Supplier Alkali Scientific Supplier No. NG2205
Three times more per case. Highly elastic comfortable feel. Powder-free with textured finger tips. 300 gloves per box, 3000 gloves per case. Thickness: Cuff: 2.4 mil Palm: 2.4 mil Fingers: 3.1 mil Strength: Tensile on spec sheet: 23 Mpa Tensile Typical Value: 32 Mp