Learn More
ADW DIABETES ULTICARE U-100 PET SYRINGES 29
SDP
Catalog No. NC3960562
Encompass
The UltiCare Pet Syringes U100 are made specifically for veterinary use. This pack of 100 29-gauge pet insulin syringes is designed for use with U-100. These easy-to-use Ulticare pet insulin needles are intended for single-use to ensure sterility. The clear markings make it easier to administer the correct dosage for dogs and cats.Features of Ulticare 09230 Vet U100 Pet Syringes:29G, 3/10 cc, 1/2" (12.7mm) needlesLatex-freeFor use with U-100 onlyEach box contains 100 UltiCare pet insulin syringes.
Catalog No. NC3960562 Supplier ADW DIABETES Supplier No. SY5751509230
The UltiCare Pet Syringes U100 are made specifically for veterinary use. This pack of 100 29-gauge pet insulin syringes is designed for use with U-100. These easy-to-use Ulticare pet insulin needles are intended for single-use to ensure sterility. The clear markings make it easier to administer the correct dosage for dogs and cats.Features of Ulticare 09230 Vet U100 Pet Syringes:29G, 3/10 cc, 1/2" (12.7mm) needlesLatex-freeFor use with U-100 onlyEach box contains 100 UltiCare pet insulin syringes.