Learn More
MSC Armstrong World Ceiling Tile, Mineral Fiber, 24 x 24 x 3/4 in, Square Lay-In
Catalog No. 502839695
Encompass
This ceiling tile offers a combination of sound absorption and blocking, sag/humidity resistance, and mold/mildew resistance. It is designed for durability, featuring resistance to water, soil, impact, scratches, and can be washed and scrubbed. It meets Class A fire performance and is suitable for clean room environments.
  • Provides sound absorption (NRC 0.70) and sound blocking (CAC 38).
  • Resists sag, humidity, mold, and mildew.
  • Disinfectable by fogging, spraying, or wiping.
  • Features high durability: water, soil, impact, scratch, wash, and scrub resistant.
  • Achieves Class A fire performance.
  • Suitable for Clean Room Class 100 environments.
  • Contains 76% recycled content.
  • Compliant with Build America, Buy America Act.
  • Includes a 30-year limited warranty against sag, mold, mildew, and bacterial growth.
Catalog No. 50-283-9695 Supplier MSC Supplier No. 89293971
This ceiling tile offers a combination of sound absorption and blocking, sag/humidity resistance, and mold/mildew resistance. It is designed for durability, featuring resistance to water, soil, impact, scratches, and can be washed and scrubbed. It meets Class A fire performance and is suitable for clean room environments.
  • Provides sound absorption (NRC 0.70) and sound blocking (CAC 38).
  • Resists sag, humidity, mold, and mildew.
  • Disinfectable by fogging, spraying, or wiping.
  • Features high durability: water, soil, impact, scratch, wash, and scrub resistant.
  • Achieves Class A fire performance.
  • Suitable for Clean Room Class 100 environments.
  • Contains 76% recycled content.
  • Compliant with Build America, Buy America Act.
  • Includes a 30-year limited warranty against sag, mold, mildew, and bacterial growth.