Learn More
DYNAMISM INC UltiMaker Sliding Block / Belt Assembly, 616 mm, S3/3/3E/2+ Connect
SDP
Catalog No. NC3979514
Encompass
This sliding block and belt assembly features a 616 mm timing belt, designed for use with a range of UltiMaker 3D printer models. It provides a complete solution for maintaining precise movement in compatible printers.
  • Complete sliding block assembly with a 616 mm timing belt.
  • Compatible with S3 (X-axis).
  • Compatible with 2+ Connect.
  • Compatible with 3.
  • Compatible with 3 Extended.
Catalog No. NC3979514 Supplier DYNAMISM INC Supplier No. 225586
This sliding block and belt assembly features a 616 mm timing belt, designed for use with a range of UltiMaker 3D printer models. It provides a complete solution for maintaining precise movement in compatible printers.
  • Complete sliding block assembly with a 616 mm timing belt.
  • Compatible with S3 (X-axis).
  • Compatible with 2+ Connect.
  • Compatible with 3.
  • Compatible with 3 Extended.