Learn More
3M™ Ultimate FX Full Facepiece Respirator Parts and Accessories
Catalog No. 19177819 Shop All 3M Products
Change view
Click to view available options
Model:
FF-400-03
FF-400-04
FF-400-06
FF-400-07
FF-400-08
FF-400-09
FF-400-11
FF-400-13
FF-400-15
FF-400-17
FF-400-18
Product Type:
Bayonet Assembly
Exhalation Valve
Head Harness Assembly
Lens Assembly
Lens Cover
Nosecup Assembly
Speaking Diaphragm Assembly
Valve Cover
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Product Type
19-177-819 FF-400-18 Lens Cover
19-177-810 FF-400-03 Lens Assembly
19-177-811 FF-400-04 Head Harness Assembly
19-177-812 FF-400-07 Exhalation Valve
19-177-813 FF-400-08 Bayonet Assembly
19-177-814 FF-400-09 Valve Cover
19-177-815 FF-400-11 Nosecup Assembly
19-177-817 FF-400-15 Lens Cover
17-623-800 FF-400-06 Head Harness Assembly
19-177-816 FF-400-13 Speaking Diaphragm Assembly
19-177-818 FF-400-17 Lens Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 19-177-819 Supplier 3M™ Supplier No. 7010385198
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement parts and accessories for use with the Ultimate FX full facepiece reusable respirators FF-401, 402, and 403.
  • Approved 3M replacement part
  • Designed for use with the 3M Ultimate FX Full Facepiece Reusable Respirators (sold separately)
  • Maintains original performance standards of facepiece
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lens Cover
For Use With (Application) Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, health and safety testing, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Ultimate FX full facepiece reusable respirator FF-401, 402 and 403
Includes 7 removable lens cover layers
Model FF-400-18
Quantity 1/Cs.