Learn More
GFS Chemicals Inc GFS Hydrochloric Acid, PPT Grade, Trace Metal Grade, 2 L
SDP
Catalog No. NC2081474
Encompass_Preferred
GFS Chemicals Inc. Ultimate Hydrochloric Acid is a high-purity, 2 L solution designed for sensitive scientific applications. Engineered to ensure low trace metal concentrations, its PPT Grade quality minimizes contamination in critical analytical procedures, making it a reliable choice for laboratories requiring exceptional purity. This product stands out due to its low trace metals, ensuring accuracy in analyses where precision is paramount.
Catalog No. NC2081474 Supplier GFS Chemicals Inc Supplier No. 72140
GFS Chemicals Inc. Ultimate Hydrochloric Acid is a high-purity, 2 L solution designed for sensitive scientific applications. Engineered to ensure low trace metal concentrations, its PPT Grade quality minimizes contamination in critical analytical procedures, making it a reliable choice for laboratories requiring exceptional purity. This product stands out due to its low trace metals, ensuring accuracy in analyses where precision is paramount.