Learn More
WELCH MATERIALS INC Ultisil MM C18/SCX, Cartridge: 5µm, 120Å, 4.6×10mm, 2pcs
Catalog No. NC3779942
Encompass_Preferred
Ultisil MM C18/SCX column features a mixed-mode stationary phase with C18 and SCX bonding, offering distinct selectivity from traditional single bonded phase columns. Featuring dual retention mechanisms of hydrophobic interaction and ion exchange, it is ideal for analyzing unknown compounds, particularly in metabolite research. It allows for the separation of highly polar compounds without the need for ion-pairing reagents, and allows for direct MS analysis.
Catalog No. NC3779942 Supplier WELCH MATERIALS INC Supplier No. H0080804032
Ultisil MM C18/SCX column features a mixed-mode stationary phase with C18 and SCX bonding, offering distinct selectivity from traditional single bonded phase columns. Featuring dual retention mechanisms of hydrophobic interaction and ion exchange, it is ideal for analyzing unknown compounds, particularly in metabolite research. It allows for the separation of highly polar compounds without the need for ion-pairing reagents, and allows for direct MS analysis.