Learn More
WELCH MATERIALS INC Ultisil XB-C18, 5μm, 10×300mm, 120Å
Supplier: WELCH MATERIALS INC H0261021103
Welch preparative column uses the same packing materials as does analytical column, to ensure the best reproducibility of analysis and preparative scale. Special packing technique and stainless modular column tube ensure the stability of column bed. The theoretical plate number for 10 μm column is greater than 35000/m; for 5 μm column, it is greater than 75000/m, and the peak symmetry is around 0.95~1.20.Ultisil XB-C18 is one of the most versatile C18 analytical columns on the market. It features advanced bonding and double-endcapping technology, delivering nearly perfect peak shapes. This column is ideal for analyzing complex matrix samples and provides excellent peak shapes for acidic and basic compounds. It is particularly effective for the separation of traditional Chinese medicine compounds.