Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Silicone Tubing, 0.25 in ID, 65 Shore A

Supplier:  Accuflow Systems ULTRA652502

Encompass_Preferred

Saint-Gobain Sani-Tech Ultra-65 is an ultra-pure, platinum-cured silicone tubing engineered for bioprocess and pharmaceutical fluid transfer. It offers ultra-low extractables and extended pump life, and is compatible with common sterilization methods used in life-science workflows, making it suitable for sterile fill, media transfer, and cell culture applications.

  • Material: platinum-cured silicone.
  • Hardness: 65 Shore A.
  • Inner diameter: 0.25 in (6.35 mm).
  • Outer diameter: 0.38 in (9.53 mm).
  • Wall thickness: 0.06 in (1.59 mm).
  • Standard length: 25 ft coil.
  • Sterilization: autoclavable; gamma irradiation up to 5.0 Mrad (50 kGy); ethylene oxide.
  • Key properties: ultra-low extractables, superior pump life.
  • Typical applications: sterile fill and transfers, drug delivery, media and buffer processing, chromatography, filtration, fermentation.

Catalog No. NC3440456


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.