Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Ultra-65, 0.25 in ID, 3/32 in Wall
Catalog No. NC3440458
Encompass_Preferred
Saint-Gobain Sani-Tech Ultra-65 silicone tubing is an ultra-pure, platinum-cured silicone tube engineered for biopharmaceutical fluid transfer. It offers low extractables, extended pump life, and broad sterilization compatibility for sterile processing and final fill operations.
  • 65 Shore A durometer hardness.
  • 0.25 in (6.35 mm) inner diameter.
  • 0.09 in (2.38 mm) wall thickness (3/32 in).
  • 0.44 in (11.11 mm) outer diameter.
  • Platinum-cured silicone material with ultra-low extractables.
  • Compatible with autoclave, gamma irradiation up to 5.0 Mrad (50 kGy), and ethylene oxide sterilization.
  • Suitable for sterile fill and transfers, drug delivery, media and buffer processing, pumping, cell culture, chromatography, final fill, and fermentation.
Catalog No. NC3440458 Supplier Accuflow Systems Supplier No. ULTRA652503
Saint-Gobain Sani-Tech Ultra-65 silicone tubing is an ultra-pure, platinum-cured silicone tube engineered for biopharmaceutical fluid transfer. It offers low extractables, extended pump life, and broad sterilization compatibility for sterile processing and final fill operations.
  • 65 Shore A durometer hardness.
  • 0.25 in (6.35 mm) inner diameter.
  • 0.09 in (2.38 mm) wall thickness (3/32 in).
  • 0.44 in (11.11 mm) outer diameter.
  • Platinum-cured silicone material with ultra-low extractables.
  • Compatible with autoclave, gamma irradiation up to 5.0 Mrad (50 kGy), and ethylene oxide sterilization.
  • Suitable for sterile fill and transfers, drug delivery, media and buffer processing, pumping, cell culture, chromatography, final fill, and fermentation.