Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain Platinum Silicone Tubing, 65 Shore A, 0.50 in ID

Catalog No. NC3440465
Encompass_Preferred

Sani-Tech Ultra-65 silicone tubing is a platinum-cured silicone tube formulated for bioprocess and pharmaceutical fluid handling. It offers firm flexibility at 65 Shore A hardness, wide temperature resistance, and standard process dimensions for sterile transfer, pumping, and fill/finish workflows.

  • Platinum-cured silicone construction.
  • 65 Shore A hardness.
  • Inner diameter 0.50 in (12.70 mm), outer diameter 0.75 in (19.05 mm).
  • Wall thickness 0.13 in (3.18 mm).
  • Typical lengths: 25 ft standard, 50 ft coil (F), 100 ft coil (H).
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Applications include sterile fill and transfers, media and buffer processing, pumping, cell culture and harvest, chromatography, and final fill.

Catalog No. NC3440465 Supplier Accuflow Systems Supplier No. ULTRA655004
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sani-Tech Ultra-65 silicone tubing is a platinum-cured silicone tube formulated for bioprocess and pharmaceutical fluid handling. It offers firm flexibility at 65 Shore A hardness, wide temperature resistance, and standard process dimensions for sterile transfer, pumping, and fill/finish workflows.

  • Platinum-cured silicone construction.
  • 65 Shore A hardness.
  • Inner diameter 0.50 in (12.70 mm), outer diameter 0.75 in (19.05 mm).
  • Wall thickness 0.13 in (3.18 mm).
  • Typical lengths: 25 ft standard, 50 ft coil (F), 100 ft coil (H).
  • Temperature range -80°F (-62°C) to 500°F (260°C).
  • Applications include sterile fill and transfers, media and buffer processing, pumping, cell culture and harvest, chromatography, and final fill.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.