Learn More
Electron Microscopy Sciences Ultra Brush
SDP
Catalog No. 5030508
Encompass_Preferred
The ULTRA BRUSH is totally different than standard camel hairbrushes. Its unique fibers are ultra-soft allowing for the cleaning of delicate surfaces of camera lenses, binoculars, telescopes and other fine optics. The ULTRA BRUSH fully retracts within the base to completely protect the brush ends. Guaranteed to be the softest lipstick style brush you can buy.
Catalog No. 50-305-08 Supplier Electron Microscopy Sciences Supplier No. 60611
The ULTRA BRUSH is totally different than standard camel hairbrushes. Its unique fibers are ultra-soft allowing for the cleaning of delicate surfaces of camera lenses, binoculars, telescopes and other fine optics. The ULTRA BRUSH fully retracts within the base to completely protect the brush ends. Guaranteed to be the softest lipstick style brush you can buy.