Learn More
Accuflow Systems Saint-Gobain Silicone Tubing, Platinum Cured, 25 ft, Biopharma Grade
Catalog No. NC3440469
Encompass_Preferred
Platinum-cured, ultra-pure silicone tubing designed for biopharmaceutical fluid transfer, peristaltic pump use, and single-use assemblies. Packaged in a certified cleanroom to minimize extractables and provide long pump life. This configuration supplies 25 ft of tubing with a 1.00 in inner diameter and 3/16 in wall thickness, suitable for demanding bioprocess environments across a wide temperature range.
  • Platinum-cured silicone construction for biocompatibility and chemical inertness.
  • Ultra-low extractables and low total organic carbon to reduce contamination risk.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C).
  • Peristaltic pump compatible, engineered for extended pump life.
  • Cleanroom packaged for single-use assemblies and sterile production workflows.
Catalog No. NC3440469 Supplier Accuflow Systems Supplier No. ULTRAC10005
Platinum-cured, ultra-pure silicone tubing designed for biopharmaceutical fluid transfer, peristaltic pump use, and single-use assemblies. Packaged in a certified cleanroom to minimize extractables and provide long pump life. This configuration supplies 25 ft of tubing with a 1.00 in inner diameter and 3/16 in wall thickness, suitable for demanding bioprocess environments across a wide temperature range.
  • Platinum-cured silicone construction for biocompatibility and chemical inertness.
  • Ultra-low extractables and low total organic carbon to reduce contamination risk.
  • Wide temperature range: -80°F (-62°C) to 500°F (260°C).
  • Peristaltic pump compatible, engineered for extended pump life.
  • Cleanroom packaged for single-use assemblies and sterile production workflows.