Learn More
Beckman Coulter ULTRA-CENTR. TUBES 45TI 25/BX
Catalog No. NC9911361
Encompass_Preferred
Thinwall Tubes, Polyallomer, Vol(g-Max): 94.0 mL, Size: 38 x 102 mm, 25/BX
Catalog No. NC9911361 Supplier Beckman Coulter Supplier No. 345775
Thinwall Tubes, Polyallomer, Vol(g-Max): 94.0 mL, Size: 38 x 102 mm, 25/BX