Learn More
Perkin Elmer US LLC Ultra Clean High Capacity Combi Filter
Catalog No. 502105222
Encompass_Preferred
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. This filter is perfect for purifying Flame Ionization Detector (FID) fuel gases, removing both moisture and hydrocarbons. Using this filter for FID hydrogen and air will produce a stable baseline, improving overall reproducibility and sensitivity. Ideal for use in combination with a zero air generator. H2O Capacity: 3.5 g HC Capacity: 6 g (as n-butane) Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: Inert carrier gas, He, H2, N2, Ar, Air Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg
Catalog No. 50-210-5222 Supplier Perkin Elmer US LLC Supplier No. N9306818
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. This filter is perfect for purifying Flame Ionization Detector (FID) fuel gases, removing both moisture and hydrocarbons. Using this filter for FID hydrogen and air will produce a stable baseline, improving overall reproducibility and sensitivity. Ideal for use in combination with a zero air generator. H2O Capacity: 3.5 g HC Capacity: 6 g (as n-butane) Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: Inert carrier gas, He, H2, N2, Ar, Air Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg