Learn More
Perkin Elmer US LLC Ultra Clean High Capacity Triple Filter, Helium-Specific
Catalog No. 502105209
Encompass_Preferred
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. This trap is packed and purged under helium. H2O Capacity: 1.8 g O2 Capacity: 75 mL HC Capacity: 4 g (as n-butane) Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: He Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg
Catalog No. 50-210-5209 Supplier Perkin Elmer US LLC Supplier No. N9306820
Add to Cart
Add to Cart
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. This trap is packed and purged under helium. H2O Capacity: 1.8 g O2 Capacity: 75 mL HC Capacity: 4 g (as n-butane) Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: He Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg