Learn More
Thermo Scientific Ultra Clean Septum for 40mL Vials
Catalog No. NC9856438 Shop All Thermo Scientific Products
Encompass
Septum designed to provide leak-free seal for 40mL vials using inert material allowing syringe needle insertion while maintaining tight seal to prevent leakage or contamination.
Catalog No. NC9856438 Supplier Thermo Scientific Supplier No. 055395
Septum designed to provide leak-free seal for 40mL vials using inert material allowing syringe needle insertion while maintaining tight seal to prevent leakage or contamination.