Learn More
Beckman Coulter Beckman Coulter Ultra-Clear open-top thinwall tube, 5 mL, 13 x 51 mm, round bottom, sterile, 48/pack
Catalog No. NC2581956
Encompass_Preferred
Beckman Coulter 5 mL Ultra-Clear open-top thinwall tubes are sterile, single-use centrifuge tubes engineered for ultracentrifugation. The ultra-clear material improves visibility of bands or pellets and the certified-free manufacturing reduces risk of contamination.
  • 5 mL nominal capacity.
  • 13 x 51 mm nominal dimensions.
  • Round bottom, thinwall construction.
  • Pre-sterilized by ethylene oxide; packaged to ISO 11135:2014 and ISO 11607:2006 standards.
  • Certified free of detectable DNA, DNase, RNase, PCR inhibitors, and endotoxins.
  • Pack size: 48 per pack.
  • Compatible with select ultracentrifuge rotors.
Catalog No. NC2581956 Supplier Beckman Coulter Supplier No. C14295
Beckman Coulter 5 mL Ultra-Clear open-top thinwall tubes are sterile, single-use centrifuge tubes engineered for ultracentrifugation. The ultra-clear material improves visibility of bands or pellets and the certified-free manufacturing reduces risk of contamination.
  • 5 mL nominal capacity.
  • 13 x 51 mm nominal dimensions.
  • Round bottom, thinwall construction.
  • Pre-sterilized by ethylene oxide; packaged to ISO 11135:2014 and ISO 11607:2006 standards.
  • Certified free of detectable DNA, DNase, RNase, PCR inhibitors, and endotoxins.
  • Pack size: 48 per pack.
  • Compatible with select ultracentrifuge rotors.