Learn More
Beckman Coulter Beckman Coulter Ultra-Clear Centrifuge Tube, 6.5 mL, Sterile, Round-Bottom (C14296)
Supplier:  Beckman Coulter C14296
Encompass_Preferred
Sterile ultra-clear 6.5 mL thinwall centrifuge tube designed for ultracentrifugation; round-bottom, open-top construction improves sample visibility and rotor compatibility. Supplied in boxes for routine laboratory workflows.
  • Provides high optical clarity for easy sample observation.
  • Thinwall polypropylene construction for improved rotor fit and performance.
  • Round-bottom, open-top design suitable for ultracentrifugation.
  • Nominal dimensions 13 x 64 mm.
  • Supplied sterile and certified free in boxes of 48.
  • Compatible with a range of ultracentrifuge rotors; verify rotor specifications before use.
Catalog No. NC2581957