Learn More
Santa Cruz Biotechnology ULTRA CRUZ MOUNTING MEDIUM
Catalog No. NC9293783
Encompass
UltraCruz mounting medium, 10mL containing 1.5ug/mL 4', 6-diamidino-2-phenylindole (DAPI) for DNA counterstaining, inhibits photobleaching of probes containing fluorescin, texas red, rhodamine and other fluorochromes
Catalog No. NC9293783 Supplier Santa Cruz Biotechnology Supplier No. SC24941
UltraCruz mounting medium, 10mL containing 1.5ug/mL 4', 6-diamidino-2-phenylindole (DAPI) for DNA counterstaining, inhibits photobleaching of probes containing fluorescin, texas red, rhodamine and other fluorochromes