Learn More
Perkin Elmer US LLC Ultra Deactivated Split/Splitless Liner with Wool
Catalog No. 502113294
Encompass_Preferred
This Ultra Deactivated Split/Splitless Liner, 4 mm, with Wool is part of a GC/MS 590/690 Capillary Injector Consumable Kit (N6550448). The kit includes an autosampler, vial locator, O-rings, liner, ferruls, septa, marathon filament, and aluminum oxide powder.
Catalog No. 50-211-3294 Supplier Perkin Elmer US LLC Supplier No. 09200624
This Ultra Deactivated Split/Splitless Liner, 4 mm, with Wool is part of a GC/MS 590/690 Capillary Injector Consumable Kit (N6550448). The kit includes an autosampler, vial locator, O-rings, liner, ferruls, septa, marathon filament, and aluminum oxide powder.