Learn More
CleanSpace Technology™ Ultra Decontamination Plug Set
Catalog No. 17800644 Shop All Cleanspace Technology Products
Change view
Click to view available options
For Use With (Equipment):
CleanSpace™ Ultra Respirator unit (PAF-0070)
Catalog No. For Use With (Equipment)
17-800-644 CleanSpace™ Ultra Respirator unit (PAF-0070)
1 options
Do not wear the respirator when the plug set is in place.
Catalog No. 17-800-644 Supplier CleanSpace Technology™ Supplier No. PAF0074
Add to Cart
Add to Cart
Do not wear the respirator when the plug set is in place.
For use with the CleanSpace™ Ultra Respirator unit (PAF-0070). Seals all the openings on the CleanSpace™ Ultra PAPR unit for storage during and after decontamination. The Plug Set is made of nylon and silicone and is safe for use in decontamination operations.
  • Used with the revolutionary CleanSpace Ultra PAPR: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and breathability
  • Easy to wash and quick drying
  • Designed for long wear in harsh environments
  • Easy and quick replacement
Specifications
Material Nylon, Silicone
For Use With (Equipment) CleanSpace™ Ultra Respirator unit (PAF-0070)
Dimensions (L x W x H) 800 x 180 x 110 mm
Weight (Metric) 100 g