Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife 3.5 mm, 45° Knife angle
SDP
Catalog No. 50363482
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.5 mm
Knife angle: 45°
Catalog No. 50-363-482 Supplier Electron Microscopy Sciences Supplier No. 35US
Add to Cart
Add to Cart
Request a Quote
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.5 mm
Knife angle: 45°