Learn More
Fine Science Tools Ultra Fine Hemostats, Stainless Steel, 12 cm
SDP
Catalog No. NC9142355
Encompass
Tip Shape: Curved/ Tips: Smooth/ Tip Width: 0.6mm/ Clamping Length: 19mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm
Catalog No. NC9142355 Supplier Fine Science Tools Supplier No. 1302112
Tip Shape: Curved/ Tips: Smooth/ Tip Width: 0.6mm/ Clamping Length: 19mm/ Alloy / Material: Stainless Steel/ Length: 12.5cm