Learn More
Thomas Scientific Microscope Slides, Glass, 25 x 75mm, 90ø Ground Edges with Safety Corners, Frosted, 1 End, 1 Side, 72/Box, 20 Boxes/Case (10 Gross)
SDP
Catalog No. 501460233
Encompass_Preferred
Frosted microscope slides. Frosted: 1 end, 1 side. 25 x 75 x 1mm. 90° ground edges. Safety corners. Pre-cleaned. 72 slides per box. Packaging: Case: 10 gross (20 boxes of 72)
Catalog No. 50-146-0233 Supplier Thomas Scientific Supplier No. 1158B88
Frosted microscope slides. Frosted: 1 end, 1 side. 25 x 75 x 1mm. 90° ground edges. Safety corners. Pre-cleaned. 72 slides per box. Packaging: Case: 10 gross (20 boxes of 72)