Learn More
Scientific Instrument Services Inc Inland 19 Ultra Pump Oil, 1 Gallon
Catalog No. NC9695368
Encompass
The Inland 19 ULTRA Pump Oil ensures that the fluid will endure thermal and chemical stresses much better than standard vacuum pump oil. The ultra-refined process eliminates nitrogen, sulfur and oxygen and the aromatic hydrocarbons, leaving a clear base stock virtually free of impurities. The excellent additive package will not volatilize under thermal stress. The antioxidant will stay in solution and bond up to four times as many radicals as other antioxidants, extending the life of your pump and fluid. 1 Gallon
Catalog No. NC9695368 Supplier Scientific Instrument Services Inc Supplier No. IN191U
The Inland 19 ULTRA Pump Oil ensures that the fluid will endure thermal and chemical stresses much better than standard vacuum pump oil. The ultra-refined process eliminates nitrogen, sulfur and oxygen and the aromatic hydrocarbons, leaving a clear base stock virtually free of impurities. The excellent additive package will not volatilize under thermal stress. The antioxidant will stay in solution and bond up to four times as many radicals as other antioxidants, extending the life of your pump and fluid. 1 Gallon