Learn More
Scientific Instrument Services Inc ULTRA GRADE 45 PUMP OIL 1LTR
Catalog No. NC9008119
Encompass
Inland 45 Pump Oil Is Recommended For Most Mass Spectrometers. It Is Compatible With Buna-N, Neoprene, And Viton Seals, And Optimizes Vacuum Pump Performance. This Oil Is Nontoxic And Noncorrosive, And It Has Low Vapor Pressure, 10-7 Torr. 1 Liter Bottle.
Catalog No. NC9008119 Supplier Scientific Instrument Services Inc Supplier No. INV45L
Inland 45 Pump Oil Is Recommended For Most Mass Spectrometers. It Is Compatible With Buna-N, Neoprene, And Viton Seals, And Optimizes Vacuum Pump Performance. This Oil Is Nontoxic And Noncorrosive, And It Has Low Vapor Pressure, 10-7 Torr. 1 Liter Bottle.