Learn More
Perkin Elmer US LLC Ultra High Capacity Oxygen Filter
Catalog No. 502105365
Encompass_Preferred
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. Oxygen is a column killer it is present even in UHP gases. Because oxygen can enter a gas line at any fitting or during gas bottle exchange, the oxygen trap should be the last connection before the gas line enters the chromatograph. O2 Capacity: 150 mL Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: Inert carrier gas, He, H2, N2, Ar Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg
Catalog No. 50-210-5365 Supplier Perkin Elmer US LLC Supplier No. N9306815
This easy to use, leak-tight cartridge system enables rapid change of exhausted cartridges without interrupting supply or system operation. Oxygen is a column killer it is present even in UHP gases. Because oxygen can enter a gas line at any fitting or during gas bottle exchange, the oxygen trap should be the last connection before the gas line enters the chromatograph. O2 Capacity: 150 mL Outlet Gas Quality (%): >99.9999 Maximum Pressure: 11 bar (160 psi) Maximum Flow: 7 L/min Usable for: Inert carrier gas, He, H2, N2, Ar Dimensions: 24 cm x 4.4 cm Weight: 0.26 kg