Learn More
VAZYME INTERNATIONAL LLC Vazyme Master Mix, 2 ×, Dye Plus, 1 mL
Catalog No. NC3979693
Encompass
This 2 × Phanta Ultra Master Mix (Dye Plus) is designed for PCR applications, offering a perfect balance of precision and speed.
  • Optimized for high-fidelity PCR.
  • Includes dye for convenient reaction monitoring and direct gel loading.
  • Provides a 2X concentration for simplified experimental setup, reducing pipetting steps.
Catalog No. NC3979693 Supplier VAZYME INTERNATIONAL LLC Supplier No. P52801
This 2 × Phanta Ultra Master Mix (Dye Plus) is designed for PCR applications, offering a perfect balance of precision and speed.
  • Optimized for high-fidelity PCR.
  • Includes dye for convenient reaction monitoring and direct gel loading.
  • Provides a 2X concentration for simplified experimental setup, reducing pipetting steps.