Learn More
Analytical Sales & Services Ultra High Pressure PEEK/SS Micro Tee for 1/32" OD Tubing
Catalog No. 5000806272
Encompass
Ultra High Pressure PEEK/SS Micro Tee for 1/32" OD Tubing
Qty/Pkg: EACH
Catalog No. 50-008-06272 Supplier Analytical Sales & Services Supplier No. 82132
Ultra High Pressure PEEK/SS Micro Tee for 1/32" OD Tubing
Qty/Pkg: EACH