Learn More
Analytical Sales & Services Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Catalog No. 5000806570
Encompass
Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Qty/Pkg: EACH
Catalog No. 50-008-06570 Supplier Analytical Sales & Services Supplier No. 83132
Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Qty/Pkg: EACH