Learn More
Analytical Sales & Services Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Catalog No. 5000805550
Encompass
Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Qty/Pkg: EACH
Catalog No. 50-008-05550 Supplier Analytical Sales & Services Supplier No. 83133
Ultra High Pressure PEEK/SS Micro Union for 1/32" OD Tubing
Qty/Pkg: EACH