Learn More
Electron Microscopy Sciences SEM Ultra High Resolution Gold on Carbon Test Specimen, Hitachi Stub
SDP
Catalog No. 5029552
Encompass_Preferred
With gold particles ranging from <1 nm to 20 nm, this test is suitable for ultra high resolution SEM tests such as field emission electron source. A magnification of at least 80,000x is required for this test.
Mount: Hitachi Stub
Catalog No. 50-295-52 Supplier Electron Microscopy Sciences Supplier No. 7951204
With gold particles ranging from <1 nm to 20 nm, this test is suitable for ultra high resolution SEM tests such as field emission electron source. A magnification of at least 80,000x is required for this test.
Mount: Hitachi Stub