Learn More
MSC Vikan Ultra Hygiene Squeegee, 400 mm, White, 1 ea
Catalog No. 502869903
Encompass
This ultra-hygienic squeegee combines ultimate hygiene with effective water removal from walls, floors and tables. The angled blade makes it easy to remove water from corners and other difficult-to-reach areas, and the splash guard ensures that liquid does not spill onto the dried surface.
  • Fully-moulded blade eliminates common harborage points and contamination traps
  • Angled blade and flexible design efficiently remove water and debris
  • Built-in splash guard prevents liquids from splashing onto dried surfaces
  • Efficiently removes liquids and dry powders from smooth surfaces
  • Provides long-lasting performance, withstanding harsh chemicals and cleaning agents
  • Available in 12 colours for use with hygienic zoning plans and 5S lean programmes
  • Contains food-safe ISCC Certified Bio-Circular Plastic
  • Compatible with all Euro threaded handles
Catalog No. 50-286-9903 Supplier MSC Supplier No. 47167481
This ultra-hygienic squeegee combines ultimate hygiene with effective water removal from walls, floors and tables. The angled blade makes it easy to remove water from corners and other difficult-to-reach areas, and the splash guard ensures that liquid does not spill onto the dried surface.
  • Fully-moulded blade eliminates common harborage points and contamination traps
  • Angled blade and flexible design efficiently remove water and debris
  • Built-in splash guard prevents liquids from splashing onto dried surfaces
  • Efficiently removes liquids and dry powders from smooth surfaces
  • Provides long-lasting performance, withstanding harsh chemicals and cleaning agents
  • Available in 12 colours for use with hygienic zoning plans and 5S lean programmes
  • Contains food-safe ISCC Certified Bio-Circular Plastic
  • Compatible with all Euro threaded handles