Learn More
Perkin Elmer US LLC Ultra Inert Splitless Liner without Wool
Catalog No. 502113296
Encompass_Preferred
This 4 mm Ultra Inert Splitless Liner without Wool features a consistent level of inertness for better sensitivity, accuracy, and reproducibility. Liners with inert wool retain non-volatiles which make trace level analysis easier.
Catalog No. 50-211-3296 Supplier Perkin Elmer US LLC Supplier No. 09200696
This 4 mm Ultra Inert Splitless Liner without Wool features a consistent level of inertness for better sensitivity, accuracy, and reproducibility. Liners with inert wool retain non-volatiles which make trace level analysis easier.