Learn More
Biolegend Ultra-LEAF™ Purified anti-mouse CD335 (NKp46)
Catalog No. NC1751727
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 1 mg
Catalog No. NC1751727 Supplier Biolegend Supplier No. 137640
Ultra-LEAF™ Purified anti-mouse CD335 (NKp46) [29A1.4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC-F; Size: 1 mg