Learn More
Biolegend Ultra-LEAF™ Purified anti-human IFN-γ
Supplier:  Biolegend 506532
Encompass_Preferred
Ultra-LEAF™ Purified anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC, Neut, FC; Size: 1 mg
Catalog No. NC1724220