Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD3
Supplier:  Biolegend 300465
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, CyTOF, IHC-F, IP, Activ, WB; Size: 5 mg
Catalog No. NC1412258