Learn More
Biolegend Ultra-LEAF™ Purified Human IgG2 Isotype Control Recombinant
Supplier:  Biolegend 403601
Encompass_Preferred
Ultra-LEAF™ Purified Human IgG2 Isotype Control Recombinant [QA16A13]; Isotype: Human IgG2, κ; Apps: FC; Size: 100 ug
Catalog No. NC1304467