Learn More
Biolegend Ultra-LEAF™ Purified Mouse IgG1, κ isotype Ctrl
Supplier:  Biolegend 400197
Encompass_Preferred
Ultra-LEAF™ Purified Mouse IgG1, κ isotype Ctrl [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse ; Apps: FC, ICFC, ICC, IHC, IP, WB, FA; Size: 5 mg
Catalog No. NC1411783