Learn More
Biolegend Ultra-LEAF™ Purified anti-human CD28 Recombinant [QA17A32]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Costim; Size: 1 mg
Supplier:  Biolegend 377304
Encompass_Preferred
Ultra-LEAF™ Purified anti-human CD28 Recombinant [QA17A32]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, Costim; Size: 1 mg
Catalog No. 50-255-1013