Learn More
Biolegend Ultra-LEAF™ Purified anti-human Glypican-4 [A21050B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICC, FC, Block; Size: 100 ug
Supplier:  Biolegend 386403
Encompass_Preferred
Ultra-LEAF™ Purified anti-human Glypican-4 [A21050B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICC, FC, Block; Size: 100 ug
Catalog No. 50-252-7682