Learn More
Biolegend Ultra-LEAFTM Purified anti-hum
Catalog No. 502722397
Encompass_Preferred
Ultra-LEAF™ Purified anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, Mouse, Rat; Apps: FC, IP, Costim; Size: 1 mg
Catalog No. 50-272-2397 Supplier Biolegend Supplier No. 313540
Ultra-LEAF™ Purified anti-human/mouse/rat CD278 (ICOS) [C398.4A]; Isotype: Armenian Hamster IgG; Reactivity: Human, Mouse, Rat; Apps: FC, IP, Costim; Size: 1 mg