Learn More
Biolegend Ultra-LEAFTM Purified anti-hum
Supplier:  Biolegend 636054
Encompass_Preferred
Ultra-LEAF™ Purified anti-human Cripto (TDGF-1) [W22247A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: Block; Size: 1 mg
Catalog No. 50-274-5573