Learn More
Biolegend Ultra-LEAFTM Purified anti-mou
Supplier:  Biolegend 137308
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse/human CD351 (Fc α/µ receptor) [TX61]; Isotype: Mouse IgG1, κ; Reactivity: Mouse, Human; Apps: FC, Block; Size: 1 mg
Catalog No. 50-272-2678