Learn More
Biolegend Ultra-LEAFTM Purified anti-mou
Supplier:  Biolegend 938904
Encompass_Preferred
Ultra-LEAF™ Purified anti-mouse CX3CL1 (Fractalkine) [A19080D]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: Neut; Size: 1 mg
Catalog No. 50-272-1604