Learn More
Biolegend Ultra-LEAFTM Purified Human Ig
Catalog No. 503107966
Encompass_Preferred
Ultra-LEAF Purified Human IgG3 Isotype Ctrl Recombinant [QA16A14] Isotype Human IgG3 Apps FC Size 100 g
Catalog No. 50-310-7966 Supplier Biolegend Supplier No. 403303
Ultra-LEAF Purified Human IgG3 Isotype Ctrl Recombinant [QA16A14] Isotype Human IgG3 Apps FC Size 100 g